Product Details

SNP ID
rs150176330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52315041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCCTGTCATCTATGCAGTCTCC[A/C]TGCTCTTCTGCTCGCTGCCGCTGCT
Phenotype
MIM: 176804
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTGER2 PubMed Links

Gene Details

Gene
PTGER2
Gene Name
prostaglandin E receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000956.3 744 Missense Mutation ATG,CTG M165L NP_000947.2

View Full Product Details