Product Details

SNP ID
rs137980277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:70832210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTTTTCTCCTGGGTCCCCATCT[C/T]GGCTTGGCGACGCCTCCGGACCCAG
Phenotype
MIM: 611300
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LARP6 PubMed Links

Gene Details

Gene
LARP6
Gene Name
La ribonucleoprotein domain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286679.1 1230 Missense Mutation AAG,GAG K256E NP_001273608.1
NM_018357.3 1230 Missense Mutation AAG,GAG K440E NP_060827.2
NM_197958.2 1230 Intron NP_932062.1

View Full Product Details