Product Details

SNP ID
rs140749690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66387416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGACGGCTCTGCAGTTAACGGGAC[C/T]AGCTCTGCGGAGTAAGTATGGGGCG
Phenotype
MIM: 176872 MIM: 610716
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP2K1 PubMed Links

Gene Details

Gene
MAP2K1
Gene Name
mitogen-activated protein kinase kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002755.3 495 Intron NP_002746.1
XM_011521783.2 495 Intron XP_011520085.1
XM_017022411.1 495 Silent Mutation ACC,ACT T23T XP_016877900.1
XM_017022412.1 495 Intron XP_016877901.1
XM_017022413.1 495 Intron XP_016877902.1
Gene
TIPIN
Gene Name
TIMELESS interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289986.1 495 Intron NP_001276915.1
NM_017858.2 495 Intron NP_060328.2
XM_005254521.2 495 Intron XP_005254578.1
XM_005254522.4 495 Intron XP_005254579.1
XM_006720594.3 495 Intron XP_006720657.1
XM_011521743.1 495 Intron XP_011520045.1
XM_017022390.1 495 Intron XP_016877879.1
XM_017022391.1 495 Intron XP_016877880.1

View Full Product Details