Product Details

SNP ID
rs148274809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43276517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCTTTGATCTCCTTGACCTCGT[C/T]GCTGCTGATGAGAACCTGGAGCTGG
Phenotype
MIM: 603805 MIM: 606776
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TGM5 PubMed Links

Gene Details

Gene
TGM5
Gene Name
transglutaminase 5
There are no transcripts associated with this gene.

Gene
TGM7
Gene Name
transglutaminase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052955.2 2074 Missense Mutation AAC,GAC N691D NP_443187.1
XM_017021903.1 2074 Missense Mutation AAC,GAC N692D XP_016877392.1

View Full Product Details