Product Details

SNP ID
rs150581104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89683891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATAAGCGAGGTACCAGGTCAGTG[A/G]CATGAATGCTCTGCTCAGTTGCCTG
Phenotype
MIM: 603866 MIM: 170290
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PEX11A PubMed Links

Gene Details

Gene
PEX11A
Gene Name
peroxisomal biogenesis factor 11 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271572.1 266 Intron NP_001258501.1
NM_001271573.1 266 UTR 5 NP_001258502.1
NM_003847.2 266 Missense Mutation GCC,GTC A77V NP_003838.1
Gene
PLIN1
Gene Name
perilipin 1
There are no transcripts associated with this gene.

Gene
WDR93
Gene Name
WD repeat domain 93
There are no transcripts associated with this gene.

View Full Product Details