Product Details

SNP ID
rs151155650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43279130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGACACTACCTCAATAGACAAG[A/T]GGGGAGGCTCCAGACAGATATCTTT
Phenotype
MIM: 606776
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TGM7 PubMed Links

Gene Details

Gene
TGM7
Gene Name
transglutaminase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052955.2 1829 Missense Mutation CAC,CTC H609L NP_443187.1
XM_017021903.1 1829 Missense Mutation CAC,CTC H610L XP_016877392.1

View Full Product Details