Product Details

SNP ID
rs138685191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66908427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTCAGGGGCACGCTGTCTGCTG[A/G]TTGATCCGGGTCCTTCTTCCTTGAC
Phenotype
MIM: 603118
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH16 PubMed Links

Gene Details

Gene
CDH16
Gene Name
cadherin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204744.1 2540 Missense Mutation CCA,TCA P797S NP_001191673.1
NM_001204745.1 2540 Missense Mutation CCA,TCA P780S NP_001191674.1
NM_001204746.1 2540 Missense Mutation CCA,TCA P722S NP_001191675.1
NM_004062.3 2540 Missense Mutation CCA,TCA P819S NP_004053.1
XM_005255770.2 2540 Missense Mutation CCA,TCA P641S XP_005255827.1
XM_011522807.1 2540 Missense Mutation CCA,TCA P756S XP_011521109.1

View Full Product Details