Product Details

SNP ID
rs138916531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:24543228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAATTTTAAAATCCCTAATCTA[C/T]CATGTTTCTGTAAAACATTTCCTTA
Phenotype
MIM: 600938
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBBP6 PubMed Links

Gene Details

Gene
RBBP6
Gene Name
RB binding protein 6, ubiquitin ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006910.4 Intron NP_008841.2
NM_018703.3 Intron NP_061173.1
NM_032626.5 Intron NP_116015.2

View Full Product Details