Product Details

SNP ID
rs144456902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11679330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTGGACCTCTCTCTGGGCCGC[C/T]AGGCTTTCAGCTCCATCCCTGCCCT
Phenotype
MIM: 603032
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNN PubMed Links

Gene Details

Gene
SNN
Gene Name
stannin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003498.5 2727 Intron NP_003489.1
XM_017023741.1 2727 Intron XP_016879230.1
XM_017023742.1 2727 Intron XP_016879231.1
Gene
TXNDC11
Gene Name
thioredoxin domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303447.1 2727 Silent Mutation CTA,CTG L941L NP_001290376.1
NM_001324022.1 2727 Silent Mutation CTA,CTG L700L NP_001310951.1
NM_001324024.1 2727 Silent Mutation CTA,CTG L462L NP_001310953.1
NM_001324025.1 2727 Silent Mutation CTA,CTG L462L NP_001310954.1
NM_015914.6 2727 Silent Mutation CTA,CTG L914L NP_056998.4
XM_011522515.2 2727 Silent Mutation CTA,CTG L954L XP_011520817.1
XM_011522516.2 2727 Silent Mutation CTA,CTG L740L XP_011520818.1
XM_017023268.1 2727 Silent Mutation CTA,CTG L953L XP_016878757.1

View Full Product Details