Product Details

SNP ID
rs145127821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30001495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGCCCAGTCCCCTGAGGCCCAAG[C/T]GGGAGAAACGGCCCCGCCTGCCCCG
Phenotype
MIM: 604567 MIM: 603365 MIM: 613199
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOC2A PubMed Links

Gene Details

Gene
DOC2A
Gene Name
double C2 domain alpha
There are no transcripts associated with this gene.

Gene
HIRIP3
Gene Name
HIRA interacting protein 3
There are no transcripts associated with this gene.

Gene
INO80E
Gene Name
INO80 complex subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304562.1 564 Intron NP_001291491.1
NM_001304563.1 564 Missense Mutation CGG,TGG R160W NP_001291492.1
NM_173618.2 564 Missense Mutation CGG,TGG R160W NP_775889.1
XM_011545809.2 564 Missense Mutation CGG,TGG R160W XP_011544111.1
XM_011545811.2 564 Missense Mutation CGG,TGG R160W XP_011544113.1
XM_011545812.2 564 Missense Mutation CGG,TGG R160W XP_011544114.1
XM_017023169.1 564 Missense Mutation CGG,TGG R160W XP_016878658.1
Gene
TAOK2
Gene Name
TAO kinase 2
There are no transcripts associated with this gene.

View Full Product Details