Product Details

SNP ID
rs145240437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:722477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGGGCAGGCTGGGGAGGCCGCC[G/T]CCTCGCGGATACCCATCCAGGCTGC
Phenotype
MIM: 614666 MIM: 610998 MIM: 611118
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC78 PubMed Links

Gene Details

Gene
CCDC78
Gene Name
coiled-coil domain containing 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031737.2 761 Intron NP_001026907.2
XM_006720838.1 761 Intron XP_006720901.1
XM_006720843.3 761 Intron XP_006720906.1
XM_011522356.1 761 Intron XP_011520658.1
XM_011522357.1 761 Intron XP_011520659.1
XM_011522358.2 761 Intron XP_011520660.1
XM_011522359.1 761 Intron XP_011520661.1
XM_011522360.1 761 Intron XP_011520662.1
XM_011522361.1 761 Intron XP_011520663.1
XM_011522362.1 761 Intron XP_011520664.1
XM_011522363.1 761 Intron XP_011520665.1
XM_011522364.1 761 Intron XP_011520666.1
XM_011522365.1 761 Intron XP_011520667.1
XM_011522366.1 761 Intron XP_011520668.1
XM_011522367.1 761 Intron XP_011520669.1
XM_011522368.1 761 Intron XP_011520670.1
XM_011522369.1 761 Intron XP_011520671.1
XM_011522370.1 761 Intron XP_011520672.1
XM_011522371.2 761 Intron XP_011520673.1
XM_017022929.1 761 Intron XP_016878418.1
XM_017022930.1 761 Intron XP_016878419.1
XM_017022931.1 761 Intron XP_016878420.1
XM_017022932.1 761 Intron XP_016878421.1
Gene
FAM173A
Gene Name
family with sequence similarity 173 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271285.1 761 Missense Mutation GCC,TCC A193S NP_001258214.1
NM_023933.2 761 Missense Mutation GCC,TCC A210S NP_076422.1
Gene
HAGHL
Gene Name
hydroxyacylglutathione hydrolase-like
There are no transcripts associated with this gene.

Gene
METRN
Gene Name
meteorin, glial cell differentiation regulator
There are no transcripts associated with this gene.

Gene
NARFL
Gene Name
nuclear prelamin A recognition factor like
There are no transcripts associated with this gene.

View Full Product Details