Product Details

SNP ID
rs146805408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85780444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTACACGTGGATCGTAGGCGCTAC[A/G]CAGTCCATCGTAAACTTGGTGTTGT
Phenotype
MIM: 604886
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMC8 PubMed Links

Gene Details

Gene
EMC8
Gene Name
ER membrane protein complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142288.1 346 Intron NP_001135760.1
NM_006067.4 346 Silent Mutation TGC,TGT C136C NP_006058.1
XM_011522813.2 346 Intron XP_011521115.1
XM_017022867.1 346 Silent Mutation TGC,TGT C84C XP_016878356.1

View Full Product Details