Product Details
- SNP ID
-
rs146883847
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:21161197 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGCTTCAGAATGGCAAAAGAGGA[A/G]CCCCAGAGTATCTCAAGGGACTTGC
- Phenotype
-
MIM: 603334
MIM: 611304
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DNAH3
PubMed Links
Gene Details
- Gene
- DNAH3
- Gene Name
- dynein axonemal heavy chain 3
There are no transcripts associated with this gene.
- Gene
- TMEM159
- Gene Name
- transmembrane protein 159
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001301769.1 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
NP_001288698.1 |
NM_001301771.1 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
NP_001288700.1 |
NM_001301773.1 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
NP_001288702.1 |
NM_001301774.1 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
NP_001288703.1 |
NM_001301775.1 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
NP_001288704.1 |
NM_020422.5 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
NP_065155.3 |
XM_005255439.4 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
XP_005255496.1 |
XM_006721066.2 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
XP_006721129.1 |
XM_006721067.3 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
XP_006721130.1 |
XM_006721069.3 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
XP_006721132.1 |
XM_017023471.1 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
XP_016878960.1 |
XM_017023472.1 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
XP_016878961.1 |
XM_017023473.1 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
XP_016878962.1 |
XM_017023474.1 |
618 |
Silent Mutation |
GAA,GAG |
E5E |
XP_016878963.1 |
View Full Product Details