Product Details

SNP ID
rs121912424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63930095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCCATGACTCGCAGCTCTGTGC[T/C]GCAGCCCTGGTAGACCTCCGAGGTG
Phenotype
MIM: 147245 MIM: 603967
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CD79B PubMed Links

Gene Details

Gene
CD79B
Gene Name
CD79b molecule
There are no transcripts associated with this gene.

Gene
SCN4A
Gene Name
sodium voltage-gated channel alpha subunit 4
There are no transcripts associated with this gene.

View Full Product Details