Product Details
- SNP ID
-
rs137917213
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:5033674 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCACCATGGGCTGAGAACAGCTGA[A/T]GGGCCTCAGGGTCTGGGCCAGGGAT
- Phenotype
-
MIM: 603060
MIM: 607883
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KIF1C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs346821] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KIF1C
- Gene Name
- kinesin family member 1C
There are no transcripts associated with this gene.
- Gene
- SLC52A1
- Gene Name
- solute carrier family 52 member 1
View Full Product Details