Product Details

SNP ID
rs138502311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40960385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTATTCTTGGTTCTGTCTTTCCAG[A/G]GCACACCGGATCTCTGCCAGCTGAG
Phenotype
MIM: 616678
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT39 PubMed Links

Gene Details

Gene
KRT39
Gene Name
keratin 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213656.3 1149 Silent Mutation GCC,GCT A371A NP_998821.3

View Full Product Details