Product Details

SNP ID
rs140430864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41873888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGACCTCAGTGATGGGCATGCC[C/T]GCGTAGATGAGCTCCTGTCCTCGCT
Phenotype
MIM: 108728
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACLY PubMed Links

Gene Details

Gene
ACLY
Gene Name
ATP citrate lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001096.2 2735 Silent Mutation GCA,GCG A855A NP_001087.2
NM_001303274.1 2735 Silent Mutation GCA,GCG A909A NP_001290203.1
NM_001303275.1 2735 Silent Mutation GCA,GCG A899A NP_001290204.1
NM_198830.1 2735 Silent Mutation GCA,GCG A845A NP_942127.1
XM_005257395.1 2735 Silent Mutation GCA,GCG A855A XP_005257452.1
XM_017024688.1 2735 Silent Mutation GCA,GCG A845A XP_016880177.1
Gene
KLHL11
Gene Name
kelch like family member 11
There are no transcripts associated with this gene.

View Full Product Details