Product Details

SNP ID
rs142347762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40184624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGGACGAGTCCCTTTACCAGG[C/G]CCTCCGAGAGGACACTCTGAGGCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6867 PubMed Links

Gene Details

Gene
MIR6867
Gene Name
microRNA 6867
There are no transcripts associated with this gene.

Gene
RAPGEFL1
Gene Name
Rap guanine nucleotide exchange factor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303533.1 7 Missense Mutation GCC,GGC A109G NP_001290462.1
NM_001303534.1 7 Missense Mutation GCC,GGC A103G NP_001290463.1
NM_016339.4 7 Missense Mutation GCC,GGC A54G NP_057423.1
XM_006721932.3 7 Missense Mutation GCC,GGC A260G XP_006721995.1
XM_006721935.3 7 Missense Mutation GCC,GGC A91G XP_006721998.1
XM_006721937.3 7 Missense Mutation GCC,GGC A54G XP_006722000.1
XM_006721938.3 7 UTR 5 XP_006722001.1

View Full Product Details