Product Details

SNP ID
rs142355604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75501812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACCTTGAGGAGGGCAGCAGGGG[C/T]GGCCGGGGGTTCTGTCTCCCCATTC
Phenotype
MIM: 612185
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASKIN2 PubMed Links

Gene Details

Gene
CASKIN2
Gene Name
CASK interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142643.2 3849 Missense Mutation ACC,GCC T1006A NP_001136115.1
NM_020753.4 3849 Missense Mutation ACC,GCC T1088A NP_065804.2
Gene
MIR6785
Gene Name
microRNA 6785
There are no transcripts associated with this gene.

Gene
TMEM94
Gene Name
transmembrane protein 94
There are no transcripts associated with this gene.

View Full Product Details