Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042471.1 | 2790 | Silent Mutation | NP_001035936.1 | ||
NM_001165958.1 | 2790 | Silent Mutation | NP_001159430.1 | ||
NM_001165959.1 | 2790 | Silent Mutation | NP_001159431.1 | ||
NM_018530.2 | 2790 | Silent Mutation | NP_061000.2 | ||
XM_011525004.1 | 2790 | Missense Mutation | XP_011523306.1 | ||
XM_011525006.1 | 2790 | Missense Mutation | XP_011523308.1 | ||
XM_011525007.2 | 2790 | Missense Mutation | XP_011523309.1 | ||
XM_011525009.1 | 2790 | Missense Mutation | XP_011523311.1 | ||
XM_011525011.1 | 2790 | Missense Mutation | XP_011523313.1 | ||
XM_011525012.1 | 2790 | Missense Mutation | XP_011523314.1 | ||
XM_011525013.1 | 2790 | Missense Mutation | XP_011523315.1 | ||
XM_011525015.1 | 2790 | Missense Mutation | XP_011523317.1 | ||
XM_017024848.1 | 2790 | Missense Mutation | XP_016880337.1 | ||
XM_017024849.1 | 2790 | Missense Mutation | XP_016880338.1 | ||
XM_017024850.1 | 2790 | Missense Mutation | XP_016880339.1 | ||
XM_017024851.1 | 2790 | Missense Mutation | XP_016880340.1 | ||
XM_017024852.1 | 2790 | Missense Mutation | XP_016880341.1 | ||
XM_017024853.1 | 2790 | Intron | XP_016880342.1 | ||
XM_017024854.1 | 2790 | Intron | XP_016880343.1 |