Product Details

SNP ID
rs143918037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81229015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGGACGATGTTTGGGGGGCGCG[C/G]GGAGCATCAGGACAGGACTCTCCCG
Phenotype
MIM: 613479
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEP131 PubMed Links

Gene Details

Gene
CEP131
Gene Name
centrosomal protein 131
There are no transcripts associated with this gene.

Gene
LOC105371925
Gene Name
uncharacterized LOC105371925
There are no transcripts associated with this gene.

Gene
TEPSIN
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144679.2 2794 Silent Mutation CCC,CCG P497P NP_653280.1
XM_005257066.2 2794 Silent Mutation CCC,CCG P775P XP_005257123.1
XM_005257067.2 2794 Silent Mutation CCC,CCG P565P XP_005257124.1
XM_006721709.3 2794 Silent Mutation CCC,CCG P567P XP_006721772.1
XM_006721712.2 2794 Silent Mutation CCC,CCG P413P XP_006721775.1
XM_011524355.1 2794 Silent Mutation CCC,CCG P777P XP_011522657.1
XM_011524356.1 2794 Silent Mutation CCC,CCG P709P XP_011522658.1
XM_011524357.1 2794 Silent Mutation CCC,CCG P560P XP_011522659.1
XM_011524358.2 2794 Silent Mutation CCC,CCG P560P XP_011522660.1
XM_017024202.1 2794 Silent Mutation CCC,CCG P608P XP_016879691.1
XM_017024203.1 2794 Silent Mutation CCC,CCG P587P XP_016879692.1
XM_017024204.1 2794 Silent Mutation CCC,CCG P560P XP_016879693.1

View Full Product Details