Product Details
- SNP ID
-
rs144459016
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:81229163 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGCTGTCCGTGCCCCCTGGGATC[C/T]GTTCAGGTCTCCACCGCCTGCTTTC
- Phenotype
-
MIM: 613479
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP131
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2048057] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEP131
- Gene Name
- centrosomal protein 131
There are no transcripts associated with this gene.
- Gene
- LOC105371925
- Gene Name
- uncharacterized LOC105371925
There are no transcripts associated with this gene.
- Gene
- TEPSIN
- Gene Name
- TEPSIN, adaptor related protein complex 4 accessory protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144679.2 |
2646 |
Missense Mutation |
CAG,CGG |
Q448R |
NP_653280.1 |
XM_005257066.2 |
2646 |
Missense Mutation |
CAG,CGG |
Q726R |
XP_005257123.1 |
XM_005257067.2 |
2646 |
Missense Mutation |
CAG,CGG |
Q516R |
XP_005257124.1 |
XM_006721709.3 |
2646 |
Missense Mutation |
CAG,CGG |
Q518R |
XP_006721772.1 |
XM_006721712.2 |
2646 |
Missense Mutation |
CAG,CGG |
Q364R |
XP_006721775.1 |
XM_011524355.1 |
2646 |
Missense Mutation |
CAG,CGG |
Q728R |
XP_011522657.1 |
XM_011524356.1 |
2646 |
Missense Mutation |
CAG,CGG |
Q660R |
XP_011522658.1 |
XM_011524357.1 |
2646 |
Missense Mutation |
CAG,CGG |
Q511R |
XP_011522659.1 |
XM_011524358.2 |
2646 |
Missense Mutation |
CAG,CGG |
Q511R |
XP_011522660.1 |
XM_017024202.1 |
2646 |
Missense Mutation |
CAG,CGG |
Q559R |
XP_016879691.1 |
XM_017024203.1 |
2646 |
Missense Mutation |
CAG,CGG |
Q538R |
XP_016879692.1 |
XM_017024204.1 |
2646 |
Missense Mutation |
CAG,CGG |
Q511R |
XP_016879693.1 |
View Full Product Details