Product Details

SNP ID
rs145210225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4941085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTTTTTGTCCGCAGACCTCGGA[C/G]CACAATGCCAGCATGGACTTTGCAG
Phenotype
MIM: 131370 MIM: 606672 MIM: 176610 MIM: 610431 MIM: 604165
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ENO3 PubMed Links

Gene Details

Gene
ENO3
Gene Name
enolase 3
There are no transcripts associated with this gene.

Gene
GP1BA
Gene Name
glycoprotein Ib platelet alpha subunit
There are no transcripts associated with this gene.

Gene
PFN1
Gene Name
profilin 1
There are no transcripts associated with this gene.

Gene
RNF167
Gene Name
ring finger protein 167
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320356.1 353 Missense Mutation GAC,GAG D31E NP_001307285.1
NM_001320357.1 353 Missense Mutation GAC,GAG D31E NP_001307286.1
NM_001320358.1 353 Missense Mutation GAC,GAG D31E NP_001307287.1
NM_001320359.1 353 Missense Mutation GAC,GAG D31E NP_001307288.1
NM_001320360.1 353 Missense Mutation GAC,GAG D31E NP_001307289.1
NM_001320361.1 353 Missense Mutation GAC,GAG D31E NP_001307290.1
NM_001320362.1 353 UTR 5 NP_001307291.1
NM_001320363.1 353 UTR 5 NP_001307292.1
NM_001320364.1 353 UTR 5 NP_001307293.1
NM_001320365.1 353 UTR 5 NP_001307294.1
NM_015528.2 353 Missense Mutation GAC,GAG D31E NP_056343.1
XM_017024427.1 353 Missense Mutation GAC,GAG D31E XP_016879916.1
XM_017024428.1 353 Intron XP_016879917.1
Gene
SLC25A11
Gene Name
solute carrier family 25 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165417.1 353 Intron NP_001158889.1
NM_001165418.1 353 Intron NP_001158890.1
NM_003562.4 353 Intron NP_003553.2

View Full Product Details