Product Details
- SNP ID
-
rs145376542
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:30834478 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATATTAAAACTGAAAATGAAGCT[C/T]CAATTGAAATTAGTAGCGACGATAG
- Phenotype
-
MIM: 609534
MIM: 614853
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATAD5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11080134] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATAD5
- Gene Name
- ATPase family, AAA domain containing 5
- Gene
- CRLF3
- Gene Name
- cytokine receptor like factor 3
There are no transcripts associated with this gene.
View Full Product Details