Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001007226.1 | 2891 | UTR 3 | NP_001007227.1 | ||
NM_001007227.1 | 2891 | UTR 3 | NP_001007228.1 | ||
NM_001007228.1 | 2891 | UTR 3 | NP_001007229.1 | ||
NM_001007229.1 | 2891 | UTR 3 | NP_001007230.1 | ||
NM_001007230.1 | 2891 | UTR 3 | NP_001007231.1 | ||
NM_003563.3 | 2891 | UTR 3 | NP_003554.1 | ||
XM_005257723.4 | 2891 | Intron | XP_005257780.1 | ||
XM_005257724.4 | 2891 | Intron | XP_005257781.1 | ||
XM_017025204.1 | 2891 | Intron | XP_016880693.1 |