Product Details

SNP ID
rs145448362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74922991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGGGGTCATGCCATCCTCGTCG[A/G]GGGCATTCAGCTCCTTTCGGGTGGC
Phenotype
MIM: 607827 MIM: 607696
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTOP2 PubMed Links

Gene Details

Gene
OTOP2
Gene Name
otopetrin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178160.2 273 Intron NP_835454.1
XM_011525479.1 273 UTR 5 XP_011523781.1
Gene
USH1G
Gene Name
USH1 protein network component sans
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282489.2 273 UTR 5 NP_001269418.1
NM_173477.4 273 Missense Mutation NP_775748.2
XM_011524296.2 273 Intron XP_011522598.1

View Full Product Details