Product Details

SNP ID
rs147194757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74951609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAAACAGCTTCACGTCGGTGTC[A/G]TACCAGACAGGGGGGTCCACATTCC
Phenotype
MIM: 605752 MIM: 607828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HID1 PubMed Links

Gene Details

Gene
HID1
Gene Name
HID1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030630.2 2472 Silent Mutation TAC,TAT Y776Y NP_085133.1
XM_005257226.1 2472 Silent Mutation TAC,TAT Y775Y XP_005257283.1
Gene
OTOP3
Gene Name
otopetrin 3
There are no transcripts associated with this gene.

View Full Product Details