Product Details

SNP ID
rs148316648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75042020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAACAGAGCCTAAATTCCTGCAC[C/T]TATTTACAGTTCAATCGACCACAAG
Phenotype
MIM: 613422
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5H PubMed Links

Gene Details

Gene
ATP5H
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003785.1 Intron NP_001003785.1
NM_006356.2 Intron NP_006347.1
Gene
KCTD2
Gene Name
potassium channel tetramerization domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015353.2 Intron NP_056168.1
XM_011524575.2 Intron XP_011522877.1

View Full Product Details