Product Details

SNP ID
rs148660107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50863557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCAGAGTGACCAAAAGGAGGC[A/G]ATGGAGAGCTGGACACTGATGAGGC
Phenotype
MIM: 605523
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TOB1 PubMed Links

Gene Details

Gene
TOB1
Gene Name
transducer of ERBB2, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243877.1 701 Missense Mutation TCG,TTG S154L NP_001230806.1
NM_001243885.1 701 Missense Mutation TCG,TTG S15L NP_001230814.1
NM_005749.3 701 Missense Mutation TCG,TTG S154L NP_005740.1
Gene
TOB1-AS1
Gene Name
TOB1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details