Product Details

SNP ID
rs150387943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8039535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCCAGCTGACACCGCCGCGGGG[A/C]GGCCACCTCCTCTTCCCCTGCTACC
Phenotype
MIM: 603697
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALOX15B PubMed Links

Gene Details

Gene
ALOX15B
Gene Name
arachidonate 15-lipoxygenase, type B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039130.1 413 Silent Mutation GGA,GGC G99G NP_001034219.1
NM_001039131.1 413 Silent Mutation GGA,GGC G99G NP_001034220.1
NM_001141.2 413 Silent Mutation GGA,GGC G99G NP_001132.2

View Full Product Details