Product Details

SNP ID
rs138574407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10923575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCCACCAGCCTGGTGTCCTCAC[A/G]GACCACGGGCCAGAGGGTGAATACC
Phenotype
MIM: 603934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf52 PubMed Links

Gene Details

Gene
C19orf52
Gene Name
chromosome 19 open reading frame 52
There are no transcripts associated with this gene.

Gene
CARM1
Gene Name
coactivator associated arginine methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199141.1 824 Intron NP_954592.1
XM_005259708.4 824 Intron XP_005259765.1
XM_011527638.2 824 Intron XP_011525940.1
Gene
YIPF2
Gene Name
Yip1 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321439.1 824 Missense Mutation CGT,TGT R252C NP_001308368.1
NM_001321440.1 824 Missense Mutation CGT,TGT R213C NP_001308369.1
NM_024029.4 824 Missense Mutation CGT,TGT R252C NP_076934.1
XM_011528270.2 824 Missense Mutation CGT,TGT R252C XP_011526572.1
XM_011528271.2 824 Missense Mutation CGT,TGT R223C XP_011526573.1

View Full Product Details