Product Details

SNP ID
rs138806414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38409187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGACTGAATGGGCCCCTGCTGCT[C/T]GACCCTCAAAGTTCTTTGCCCTATA
Phenotype
MIM: 607320 MIM: 609293
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM98C PubMed Links

Gene Details

Gene
FAM98C
Gene Name
family with sequence similarity 98 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174905.3 2987 Intron NP_777565.3
XM_017026354.1 2987 Intron XP_016881843.1
Gene
RASGRP4
Gene Name
RAS guanyl releasing protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146202.1 2987 UTR 3 NP_001139674.1
NM_001146203.1 2987 UTR 3 NP_001139675.1
NM_001146204.1 2987 UTR 3 NP_001139676.1
NM_001146205.1 2987 UTR 3 NP_001139677.1
NM_001146206.1 2987 UTR 3 NP_001139678.1
NM_001146207.1 2987 UTR 3 NP_001139679.1
NM_170604.2 2987 UTR 3 NP_733749.1
Gene
SPRED3
Gene Name
sprouty related EVH1 domain containing 3
There are no transcripts associated with this gene.

View Full Product Details