Product Details

SNP ID
rs139711961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53914546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTCAGAATATTTTCTTGAACC[A/G]GAGATCAATTTGGTGACGGAAAACA
Phenotype
MIM: 606899 MIM: 176980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNG7 PubMed Links

Gene Details

Gene
CACNG7
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031896.4 365 Silent Mutation CCA,CCG P81P NP_114102.2
XM_005259124.3 365 Silent Mutation CCA,CCG P81P XP_005259181.1
XM_017027093.1 365 Silent Mutation CCA,CCG P81P XP_016882582.1
Gene
PRKCG
Gene Name
protein kinase C gamma
There are no transcripts associated with this gene.

View Full Product Details