Product Details

SNP ID
rs140841032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51142455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAACCGGAAGGATTACTCGCTGA[C/T]GATGCAGAGTTCCGTGACCGTGCAA
Phenotype
MIM: 604410 MIM: 605640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIGLEC7 PubMed Links

Gene Details

Gene
SIGLEC7
Gene Name
sialic acid binding Ig like lectin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277201.1 155 Missense Mutation ACG,ATG T29M NP_001264130.1
NM_014385.3 155 Missense Mutation ACG,ATG T29M NP_055200.1
NM_016543.3 155 Missense Mutation ACG,ATG T29M NP_057627.2
XM_011526721.2 155 Missense Mutation ACG,ATG T29M XP_011525023.1
Gene
SIGLEC9
Gene Name
sialic acid binding Ig like lectin 9
There are no transcripts associated with this gene.

View Full Product Details