Product Details
- SNP ID
-
rs141881690
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:54336850 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTCAGCGCCCTCACCTGAGACCA[C/T]GAGCTCCAGGGGCTCACTGGGGTGA
- Phenotype
-
MIM: 607517
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LILRA4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138326444] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LILRA4
- Gene Name
- leukocyte immunoglobulin like receptor A4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012276.4 |
1322 |
Missense Mutation |
ATG,GTG |
M416V |
NP_036408.4 |
View Full Product Details