Product Details

SNP ID
rs142027939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49061437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTCGAATCCATCGCCAGCCCAC[A/G]CGGCCCTGGGCATCAGCGGTCAATG
Phenotype
MIM: 608826 MIM: 176268 MIM: 162662
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CGB7 PubMed Links

Gene Details

Gene
CGB7
Gene Name
chorionic gonadotropin beta subunit 7
There are no transcripts associated with this gene.

Gene
KCNA7
Gene Name
potassium voltage-gated channel subfamily A member 7
There are no transcripts associated with this gene.

Gene
NTF4
Gene Name
neurotrophin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006179.4 823 Silent Mutation CGC,CGT R187R NP_006170.1
XM_005258962.3 823 Silent Mutation CGC,CGT R187R XP_005259019.1
XM_006723232.3 823 Intron XP_006723295.1
XM_011527008.2 823 Intron XP_011525310.1
XM_011527009.2 823 Intron XP_011525311.1
XM_011527010.2 823 Intron XP_011525312.1

View Full Product Details