Product Details

SNP ID
rs142171943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6752737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGGTCAGGGCCTCCCTCAGCTC[C/T]GGGTTAGGCCGGAATCCTGGAAGCA
Phenotype
MIM: 604721 MIM: 604504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH2D3A PubMed Links

Gene Details

Gene
SH2D3A
Gene Name
SH2 domain containing 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005490.2 1458 Silent Mutation CCA,CCG P529P NP_005481.2
XM_005259467.1 1458 Silent Mutation CCA,CCG P558P XP_005259524.1
XM_005259472.2 1458 Silent Mutation CCA,CCG P436P XP_005259529.1
XM_006722602.1 1458 Silent Mutation CCA,CCG P438P XP_006722665.1
XM_011527608.2 1458 Silent Mutation CCA,CCG P463P XP_011525910.1
XM_011527609.2 1458 Silent Mutation CCA,CCG P463P XP_011525911.1
XM_017026123.1 1458 Intron XP_016881612.1
XM_017026124.1 1458 Intron XP_016881613.1
Gene
TRIP10
Gene Name
thyroid hormone receptor interactor 10
There are no transcripts associated with this gene.

View Full Product Details