Product Details

SNP ID
rs142175017
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33379661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACAGTCCATTAGCACTGAAAATA[C/T]GACAGCAGATGGCGACAATGCAGGA
Phenotype
MIM: 138972 MIM: 613230
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CEBPG PubMed Links

Gene Details

Gene
CEBPG
Gene Name
CCAAT/enhancer binding protein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252296.1 608 Missense Mutation ACG,ATG T141M NP_001239225.1
NM_001806.3 608 Missense Mutation ACG,ATG T141M NP_001797.1
Gene
PEPD
Gene Name
peptidase D
There are no transcripts associated with this gene.

View Full Product Details