Product Details

SNP ID
rs142427159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6307168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGCTTACCAGATGCTGTGCAGA[C/T]AGAAGAAATGAATCCTCTGCCAGAA
Phenotype
MIM: 613491
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACER1 PubMed Links

Gene Details

Gene
ACER1
Gene Name
alkaline ceramidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133492.2 689 Missense Mutation TAT,TGT Y204C NP_597999.1
XM_011527673.2 689 Missense Mutation TAT,TGT Y161C XP_011525975.1

View Full Product Details