Product Details

SNP ID
rs143812338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55620921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTAGGCAGTAAGCTGTTTGCTT[C/T]GAACTGGGAGTGGAGGAAATTAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF784 PubMed Links

Gene Details

Gene
ZNF784
Gene Name
zinc finger protein 784
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203374.1 1816 UTR 3 NP_976308.1
Gene
ZNF865
Gene Name
zinc finger protein 865
There are no transcripts associated with this gene.

View Full Product Details