Product Details

SNP ID
rs144013891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42309642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCAGCCTGAGAAACCTGGTCGT[A/G]TCCGTCGTCGGAAGACTAGGCGGGA
Phenotype
MIM: 603074
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAFAH1B3 PubMed Links

Gene Details

Gene
PAFAH1B3
Gene Name
platelet activating factor acetylhydrolase 1b catalytic subunit 3
There are no transcripts associated with this gene.

Gene
PRR19
Gene Name
proline rich 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199285.2 148 Missense Mutation ATC,GTC I20V NP_954979.2
XM_005258779.4 148 Missense Mutation ATC,GTC I72V XP_005258836.2
XM_005258780.3 148 Missense Mutation ATC,GTC I20V XP_005258837.1
XM_006723154.3 148 Missense Mutation ATC,GTC I20V XP_006723217.1
XM_011526787.1 148 Missense Mutation ATC,GTC I20V XP_011525089.1
XM_011526788.1 148 Missense Mutation ATC,GTC I20V XP_011525090.1
XM_011526789.2 148 Missense Mutation ATC,GTC I20V XP_011525091.1
XM_011526790.2 148 Missense Mutation ATC,GTC I20V XP_011525092.1
Gene
TMEM145
Gene Name
transmembrane protein 145
There are no transcripts associated with this gene.

View Full Product Details