Product Details

SNP ID
rs144297627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57961539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCGAGCTGTTGTCTTTCCTCAGC[C/T]TGTGCCAGTCGACTGGAGAATGATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf18 PubMed Links

Gene Details

Gene
C19orf18
Gene Name
chromosome 19 open reading frame 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152474.4 486 Silent Mutation CAA,CAG Q128Q NP_689687.1

View Full Product Details