Product Details

SNP ID
rs145337836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3765230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGGCCCTCAAGGACCCCGACGT[A/G]TGCACAGATCCTGTCCAGCTCACCA
Phenotype
MIM: 604262 MIM: 611858 MIM: 610362
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APBA3 PubMed Links

Gene Details

Gene
APBA3
Gene Name
amyloid beta precursor protein binding family A member 3
There are no transcripts associated with this gene.

Gene
MRPL54
Gene Name
mitochondrial ribosomal protein L54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172251.2 217 Silent Mutation GTA,GTG V61V NP_758455.1
Gene
RAX2
Gene Name
retina and anterior neural fold homeobox 2
There are no transcripts associated with this gene.

View Full Product Details