Product Details

SNP ID
rs145420428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9102966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTGGGTGAGGCAGCCTGAGTAC[A/G]TGATGCTCCGATTCTGAGCTTGGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1M1 PubMed Links

Gene Details

Gene
OR1M1
Gene Name
olfactory receptor family 1 subfamily M member 1
There are no transcripts associated with this gene.

Gene
OR7G2
Gene Name
olfactory receptor family 7 subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005193.1 341 Missense Mutation ACG,ATG T114M NP_001005193.1

View Full Product Details