Product Details

SNP ID
rs145749824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11296360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGTTATCTTTACTCAATGGCTT[C/T]GTGGCTGTAAGTATGTCACAATCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSPC102 PubMed Links

Gene Details

Gene
HSPC102
Gene Name
uncharacterized HSPC102
There are no transcripts associated with this gene.

Gene
TSPAN16
Gene Name
tetraspanin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282509.1 222 Silent Mutation TTC,TTT F21F NP_001269438.1
NM_001282510.1 222 Silent Mutation TTC,TTT F21F NP_001269439.1
NM_012466.3 222 Silent Mutation TTC,TTT F21F NP_036598.1
XM_011527901.1 222 Silent Mutation TTC,TTT F21F XP_011526203.1
XM_011527902.1 222 Silent Mutation TTC,TTT F21F XP_011526204.1
XM_011527903.2 222 Intron XP_011526205.1

View Full Product Details