Product Details

SNP ID
rs146067641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:305939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGGAGGCTCCCTGCCTTGGTAC[C/T]GGGGGAGGACAGCAAAGGGGATGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929060 PubMed Links

Gene Details

Gene
LOC101929060
Gene Name
uncharacterized LOC101929060
There are no transcripts associated with this gene.

Gene
MIER2
Gene Name
MIER family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005259582.4 2982 UTR 3 XP_005259639.1
XM_006722769.3 2982 UTR 3 XP_006722832.1
XM_011528072.2 2982 UTR 3 XP_011526374.1
XM_011528073.1 2982 UTR 3 XP_011526375.1
XM_011528074.2 2982 UTR 3 XP_011526376.1
XM_011528075.2 2982 UTR 3 XP_011526377.1
XM_011528077.2 2982 UTR 3 XP_011526379.1
XM_011528081.2 2982 UTR 3 XP_011526383.1
XM_011528082.2 2982 UTR 3 XP_011526384.1

View Full Product Details