Product Details

SNP ID
rs146439716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39826247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCACAATATCGGTTGCTGTAG[C/T]GGTAGGTCCGGTTGTCACTGGAGGA
Phenotype
MIM: 604556 MIM: 134795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYRK1B PubMed Links

Gene Details

Gene
DYRK1B
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004714.2 1679 Missense Mutation CAC,CGC H484R NP_004705.1
NM_006483.2 1679 Missense Mutation CAC,CGC H444R NP_006474.1
NM_006484.2 1679 Missense Mutation CAC,CGC H456R NP_006475.1
XM_005259398.4 1679 Missense Mutation CAC,CGC H484R XP_005259455.1
Gene
FBL
Gene Name
fibrillarin
There are no transcripts associated with this gene.

Gene
MIR6719
Gene Name
microRNA 6719
There are no transcripts associated with this gene.

View Full Product Details