Product Details

SNP ID
rs146831669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12973466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTCATTGCTCCTCTCTCTACAT[C/G]CCTGGCTCGGACCCCACCCCACTAG
Phenotype
MIM: 609068
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DAND5 PubMed Links

Gene Details

Gene
DAND5
Gene Name
DAN domain BMP antagonist family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152654.2 Intron NP_689867.1

View Full Product Details