Product Details

SNP ID
rs146974463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10923563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGCAGCACTGTGGCCACCAGCC[G/T]GGTGTCCTCACGGACCACGGGCCAG
Phenotype
MIM: 603934
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C19orf52 PubMed Links

Gene Details

Gene
C19orf52
Gene Name
chromosome 19 open reading frame 52
There are no transcripts associated with this gene.

Gene
CARM1
Gene Name
coactivator associated arginine methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199141.1 836 Intron NP_954592.1
XM_005259708.4 836 Intron XP_005259765.1
XM_011527638.2 836 Intron XP_011525940.1
Gene
YIPF2
Gene Name
Yip1 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321439.1 836 Silent Mutation AGG,CGG R256R NP_001308368.1
NM_001321440.1 836 Silent Mutation AGG,CGG R217R NP_001308369.1
NM_024029.4 836 Silent Mutation AGG,CGG R256R NP_076934.1
XM_011528270.2 836 Silent Mutation AGG,CGG R256R XP_011526572.1
XM_011528271.2 836 Silent Mutation AGG,CGG R227R XP_011526573.1

View Full Product Details