Product Details
- SNP ID
-
hCV165460842
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:8731226 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATGTACTTCCTGCTCAGCCAGCT[C/G]TCCCTGTTTGACATTGGCTGTCCCA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OR2Z1
PubMed Links
Gene Details
- Gene
- OR2Z1
- Gene Name
- olfactory receptor family 2 subfamily Z member 1
There are no transcripts associated with this gene.
View Full Product Details